siRNA Details
Gene Symbol: NAPSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002798.1 | 1, 2 | 87 | |||
GenBank mRNA | AF090387.1 | - | 1, 2 | 94 | ||
AF098485.1 | - | 1, 2 | 94 | |||
AF200344.1 | - | 1, 2 | 95 | |||
AK300884.1 | - | 1, 2 | 98 | |||
AK310134.1 | - | 1, 2 | 105 | |||
BC085008.1 | - | 1, 2 | 40 | |||
BC111705.1 | - | 1, 2 | 84 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
