siRNA Details
Gene Symbol: LRRC4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080457.1 | NP_001073926.1 | 3 | 1396 | ||
XM_005259429.4 | XP_005259486.1 | 3 | 1575 | |||
XM_006723506.3 | XP_006723569.1 | 4 | 1677 | |||
XM_011527520.2 | XP_011525822.1 | 3 | 2823 | |||
GenBank mRNA | AL137451.1 | - | 1 | 616 | ||
BC019687.3 | - | 1 | 640 | |||
BC032460.1 | - | 2 | 1271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
