siRNA Details
Gene Symbol: NPHP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000998.1 | XP_016856487.1 | 26 | 3909 | ||
GenBank mRNA | AK127333.1 | - | 3 | 2297 | ||
BC050076.1 | - | 2 | 2195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
