siRNA Details
Gene Symbol: ZSWIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023072.2 | NP_075560.2 | 12 | 2236 | ||
XM_005260023.3 | XP_005260080.1 | 13 | 2398 | |||
XM_017027153.1 | XP_016882642.1 | 13 | 2669 | |||
XM_017027154.1 | XP_016882643.1 | 13 | 2601 | |||
XM_017027155.1 | XP_016882644.1 | 12 | 2502 | |||
XM_017027156.1 | XP_016882645.1 | 12 | 2173 | |||
GenBank mRNA | AK022283.1 | - | 1 | 19 | ||
AK024452.1 | - | 12 | 2236 | |||
AK299057.1 | - | 11 | 1774 | |||
BG764264.1 | - | 4 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
