siRNA Details
Gene Symbol: ZSWIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023072.2 | NP_075560.2 | 3, 4 | 895 | ||
XM_005260023.3 | XP_005260080.1 | 3, 4 | 706 | |||
XM_017027153.1 | XP_016882642.1 | 3, 4 | 929 | |||
XM_017027154.1 | XP_016882643.1 | 3, 4 | 930 | |||
XM_017027155.1 | XP_016882644.1 | 3, 4 | 930 | |||
XM_017027156.1 | XP_016882645.1 | 2, 3 | 433 | |||
XM_017027157.1 | XP_016882646.1 | 3, 4 | 931 | |||
GenBank mRNA | AK024452.1 | - | 3, 4 | 895 | ||
AK299057.1 | - | 1, 2 | 82 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
