siRNA Details
Gene Symbol: ZSWIM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023072.2 | NP_075560.2 | 13 | 3525 | ||
XM_005260023.3 | XP_005260080.1 | 14 | 3687 | |||
XM_017027153.1 | XP_016882642.1 | 14 | 3958 | |||
XM_017027154.1 | XP_016882643.1 | 14 | 3890 | |||
XM_017027155.1 | XP_016882644.1 | 13 | 3791 | |||
XM_017027156.1 | XP_016882645.1 | 13 | 3462 | |||
GenBank mRNA | AK022283.1 | - | 2 | 1308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
