siRNA Details
Gene Symbol: DCAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138353.2 | NP_612362.2 | 12 | 1732 | ||
GenBank mRNA | AK093072.1 | - | 12 | 1749 | ||
BC002926.2 | - | 10 | 1486 | |||
BC013280.2 | - | 10 | 1487 | |||
BC018894.2 | - | 10 | 1486 | |||
BC018895.2 | - | 10 | 1486 | |||
BC080575.1 | - | 12 | 1732 | 6580131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
