siRNA Details
Gene Symbol: DCAF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138353.2 | NP_612362.2 | 9 | 1354 | ||
GenBank mRNA | AK093072.1 | - | 9 | 1371 | ||
BC002926.2 | - | 7 | 1108 | |||
BC013280.2 | - | 7 | 1109 | |||
BC018894.2 | - | 7 | 1108 | |||
BC018895.2 | - | 7 | 1108 | |||
BC080575.1 | - | 9 | 1354 | 6580131 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
