siRNA Details
Gene Symbol: CNGA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037329.3 | NP_001032406.1 | 3 | 347 | ||
XM_017017217.1 | XP_016872706.1 | 3 | 347 | |||
XM_017017218.1 | XP_016872707.1 | 3 | 254 | |||
XM_017017219.1 | XP_016872708.1 | 3 | 255 | |||
GenBank mRNA | AK122736.1 | - | 3 | 340 | ||
AK302551.1 | - | 3 | 263 | |||
BC040277.1 | - | 3 | 286 | |||
DB238337.1 | - | 3 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
