siRNA Details
Gene Symbol: EFR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323553.1 | NP_001310482.1 | 5, 6 | 793 | ||
NM_001323554.1 | NP_001310483.1 | 5, 6 | 1732 | |||
NM_001323555.1 | NP_001310484.1 | 5, 6 | 1801 | |||
NM_001323556.1 | NP_001310485.1 | 5, 6 | 574 | |||
NM_001323557.1 | NP_001310486.1 | 5, 6 | 643 | |||
NM_001323558.1 | NP_001310487.1 | 5, 6 | 724 | |||
NM_015137.5 | NP_055952.2 | 5, 6 | 724 | |||
NR_136615.1 | 5, 6 | 793 | ||||
NR_136616.1 | 5, 6 | 724 | ||||
GenBank mRNA | AK307433.1 | - | 5, 6 | 675 | ||
BC071611.1 | - | 5, 6 | 701 | |||
BC108667.1 | - | 5, 6 | 692 | |||
BC131486.1 | - | 1, 2 | 59 | |||
BC152442.1 | - | 5, 6 | 692 | 8862567 | ||
CR749243.1 | - | 5, 6 | 534 | |||
D63477.1 | - | 5, 6 | 692 | |||
DB451954.2 | - | 5, 6 | 703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
