siRNA Details
Gene Symbol: KDM6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080424.1 | NP_001073893.1 | 22 | 6000 | ||
XM_005256549.3 | XP_005256606.1 | 23 | 6152 | |||
XM_005256550.4 | XP_005256607.1 | 22 | 5994 | |||
XM_005256551.3 | XP_005256608.1 | 22 | 6033 | |||
XM_005256552.4 | XP_005256609.1 | 22 | 7006 | |||
XM_006721483.3 | XP_006721546.1 | 23 | 6097 | |||
XM_011523750.2 | XP_011522052.1 | 23 | 7357 | |||
XM_011523752.2 | XP_011522054.1 | 22 | 5997 | |||
XM_017024381.1 | XP_016879870.1 | 23 | 7326 | |||
GenBank mRNA | AB002344.2 | - | 22 | 6005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
