siRNA Details
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 16 | 3450 | ||
NR_027409.1 | 24 | 5712 | ||||
XM_006720440.2 | XP_006720503.1 | 16 | 3415 | |||
XM_017022018.1 | XP_016877507.1 | 20 | 5846 | |||
XM_017022019.1 | XP_016877508.1 | 19 | 5542 | |||
XM_017022020.1 | XP_016877509.1 | 14 | 3537 | |||
GenBank mRNA | AB020662.1 | - | 15 | 3430 | ||
AF204231.1 | - | 15 | 3767 | |||
AL117569.1 | - | 2 | 1757 | |||
AL137686.1 | - | 15 | 3170 | |||
BC064341.1 | - | 1 | 882 | |||
BC150329.1 | - | 15 | 3441 | |||
BC152410.1 | - | 15 | 3430 | |||
BX648160.1 | - | 23 | 5666 | |||
BX648575.1 | - | 23 | 5419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
