siRNA Details
Gene Symbol: KIAA0907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014949.3 | NP_055764.2 | 7 | 939 | ||
GenBank mRNA | AB020714.1 | - | 7 | 896 | ||
AK225714.1 | - | 6 | 746 | |||
AK289902.1 | - | 7 | 882 | |||
AY112680.1 | - | 7 | 870 | |||
AY112681.1 | - | 7 | 870 | |||
AY112682.1 | - | 7 | 870 | |||
BC062637.1 | - | 7 | 894 | 5497581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
