siRNA Details
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015506.2 | NP_056321.2 | 4 | 1041 | ||
XM_005270724.4 | XP_005270781.1 | 3 | 800 | |||
XM_011541204.2 | XP_011539506.1 | 4 | 950 | |||
GenBank mRNA | AL080062.1 | - | 4 | 912 | ||
BC006122.1 | - | 4 | 884 | 3535453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
