siRNA Details
Gene Symbol: PABPC1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001124756.2 | NP_001118228.1 | 12 | 1718 | ||
NR_134983.1 | 11 | 1569 | ||||
NR_134984.1 | 3 | 1814 | ||||
NR_134985.1 | 3 | 1814 | ||||
NR_134986.1 | 3 | 1814 | ||||
NR_134987.1 | 3 | 1814 | ||||
GenBank mRNA | AK026760.1 | - | 3 | 1197 | ||
AK124047.1 | - | 3 | 1814 | |||
BC068104.1 | - | 3 | 998 | |||
BM832739.1 | - | 3 | 275 | |||
BX451955.2 | - | 5 | 675 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
