siRNA Details
Gene Symbol: LPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301860.1 | NP_001288789.1 | 16, 17 | 2197 | ||
NR_126051.1 | 16, 17 | 2192 | ||||
XM_017028020.1 | XP_016883509.1 | 16, 17 | 2192 | |||
GenBank mRNA | BC140806.1 | - | 16, 17 | 2078 | 9021674 | |
BX505800.1 | - | 5, 6 | 463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
