siRNA Details
Gene Symbol: LPIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301860.1 | NP_001288789.1 | 9 | 1481 | ||
NR_126051.1 | 9 | 1481 | ||||
XM_005260516.2 | XP_005260573.1 | 9 | 1479 | |||
XM_006723863.2 | XP_006723926.1 | 9 | 1479 | |||
XM_011528996.1 | XP_011527298.1 | 9 | 1479 | |||
XM_011528997.2 | XP_011527299.1 | 10 | 1685 | |||
XM_011528998.2 | XP_011527300.1 | 9 | 1502 | |||
XM_011528999.2 | XP_011527301.1 | 10 | 1632 | |||
XM_011529000.1 | XP_011527302.1 | 9 | 1476 | |||
XM_011529001.1 | XP_011527303.1 | 9 | 1479 | |||
XM_011529002.2 | XP_011527304.1 | 8 | 1281 | |||
XM_011529003.1 | XP_011527305.1 | 9 | 1479 | |||
XM_011529004.1 | XP_011527306.1 | 9 | 1479 | |||
XM_011529005.2 | XP_011527307.1 | 9 | 1479 | |||
XM_011529006.2 | XP_011527308.1 | 9 | 1479 | |||
XM_017028020.1 | XP_016883509.1 | 9 | 1476 | |||
GenBank mRNA | BC140806.1 | - | 9 | 1362 | 9021674 | |
BX647082.1 | - | 9 | 1438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
