siRNA Details
Gene Symbol: MTUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033602.2 | NP_001028774.2 | 5 | 2980 | ||
XM_011535019.2 | XP_011533321.1 | 8 | 3565 | |||
XM_011535020.2 | XP_011533322.1 | 7 | 3505 | |||
XM_011535021.2 | XP_011533323.1 | 7 | 3502 | |||
XM_011535022.2 | XP_011533324.1 | 5 | 630 | |||
XM_017020500.1 | XP_016875989.1 | 8 | 3565 | |||
XM_017020502.1 | XP_016875991.1 | 8 | 3513 | |||
GenBank mRNA | AB018317.2 | - | 5 | 2980 | ||
BC150245.1 | - | 5 | 3001 | 9007469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
