siRNA Details
Gene Symbol: XPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022459.4 | NP_071904.4 | 21 | 3171 | ||
XM_005266502.1 | XP_005266559.1 | 22 | 3326 | |||
XM_005266504.3 | XP_005266561.1 | 15 | 2698 | |||
XM_011535197.1 | XP_011533499.1 | 22 | 3335 | |||
XM_011535198.1 | XP_011533500.1 | 20 | 3013 | |||
XM_011535199.1 | XP_011533501.1 | 19 | 2976 | |||
XM_011535200.2 | XP_011533502.1 | 19 | 4426 | |||
XM_017020704.1 | XP_016876193.1 | 17 | 2967 | |||
XM_017020705.1 | XP_016876194.1 | 18 | 4203 | |||
XM_017020706.1 | XP_016876195.1 | 15 | 2322 | |||
GenBank mRNA | AB051508.1 | - | 21 | 3134 | ||
AK023108.1 | - | 13 | 2127 | |||
AK057924.1 | - | 21 | 3141 | |||
AL831819.2 | - | 4 | 474 | |||
BC042504.1 | - | 18 | 2707 | |||
BC113571.1 | - | 20 | 3074 | 8322623 | ||
BC113573.1 | - | 20 | 3074 | 8322625 | ||
BX647843.1 | - | 12 | 1943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
