siRNA Details
Gene Symbol: XPO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022459.4 | NP_071904.4 | 23 | 3695 | ||
XM_005266502.1 | XP_005266559.1 | 24 | 3850 | |||
XM_005266503.3 | XP_005266560.1 | 20 | 3353 | |||
XM_005266504.3 | XP_005266561.1 | 17 | 3222 | |||
XM_011535197.1 | XP_011533499.1 | 24 | 3859 | |||
XM_011535198.1 | XP_011533500.1 | 22 | 3537 | |||
XM_011535199.1 | XP_011533501.1 | 21 | 3500 | |||
XM_011535200.2 | XP_011533502.1 | 21 | 4950 | |||
XM_017020704.1 | XP_016876193.1 | 19 | 3491 | |||
XM_017020705.1 | XP_016876194.1 | 20 | 4727 | |||
XM_017020706.1 | XP_016876195.1 | 17 | 2846 | |||
GenBank mRNA | AB051508.1 | - | 23 | 3658 | ||
AL831819.2 | - | 6 | 998 | |||
BX647843.1 | - | 14 | 2467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
