siRNA Details
Gene Symbol: C12orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009894.2 | NP_001009894.2 | 5 | 711 | ||
GenBank mRNA | AK095477.1 | - | 5 | 711 | ||
AK311222.1 | - | 4 | 579 | |||
AL137488.1 | - | 4 | 2619 | |||
BC035136.2 | - | 4 | 548 | |||
BC058014.1 | - | 4 | 576 | |||
BC092422.1 | - | 5 | 653 | 30386540 | ||
BX537868.1 | - | 5 | 683 | |||
BX640968.1 | - | 5 | 686 | |||
BX648026.1 | - | 4 | 547 | |||
CR936776.1 | - | 5 | 700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
