siRNA Details
Gene Symbol: TTC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015351.1 | NP_056166.1 | 1 | 728 | ||
GenBank mRNA | BC047950.1 | - | 1 | 550 | ||
BX407741.2 | - | 1 | 555 | |||
D86980.1 | - | 1 | 728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
