siRNA Details
Gene Symbol: TTC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015351.1 | NP_056166.1 | 3 | 2140 | ||
GenBank mRNA | BC047950.1 | - | 3 | 1962 | ||
D86980.1 | - | 3 | 2140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
