siRNA Details
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009941.2 | NP_001009941.1 | 4 | 1139 | ||
NM_001009943.2 | NP_001009943.1 | 4 | 1139 | |||
NM_019046.2 | NP_061919.1 | 4 | 1139 | |||
GenBank mRNA | AW630443.1 | - | 3 | 218 | ||
BC062346.1 | - | 4 | 625 | 3845142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
