siRNA Details
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009941.2 | NP_001009941.1 | 5 | 1320 | ||
NM_001009943.2 | NP_001009943.1 | 5 | 1320 | |||
NM_019046.2 | NP_061919.1 | 5 | 1320 | |||
GenBank mRNA | AL137614.1 | - | 2 | 175 | ||
BC062346.1 | - | 5 | 806 | 3845142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
