siRNA Details
Gene Symbol: ANKRD16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019046.2 | NP_061919.1 | 8 | 2051 | ||
GenBank mRNA | AK093489.1 | - | 1 | 1362 | ||
AL137614.1 | - | 5 | 906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
