siRNA Details
Gene Symbol: LRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002334.3 | NP_002325.2 | 2 | 433 | ||
XM_017017734.1 | XP_016873223.1 | 2 | 350 | |||
GenBank mRNA | AB084910.1 | - | 2 | 433 | ||
AB231861.1 | - | 2 | 336 | |||
BC136667.1 | - | 2 | 354 | 9020657 | ||
BC136668.1 | - | 2 | 354 | 9020658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
