siRNA Details
Gene Symbol: LRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002334.3 | NP_002325.2 | 23 | 3464 | ||
XM_011520103.2 | XP_011518405.1 | 17 | 2534 | |||
XM_011520104.2 | XP_011518406.1 | 8 | 1001 | |||
XM_017017734.1 | XP_016873223.1 | 23 | 3381 | |||
GenBank mRNA | AB011540.1 | - | 15 | 2234 | ||
AB084910.1 | - | 23 | 3464 | |||
AB231861.1 | - | 23 | 3367 | |||
BC136667.1 | - | 23 | 3385 | 9020657 | ||
BC136668.1 | - | 23 | 3385 | 9020658 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
