siRNA Details
Gene Symbol: FHOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281739.2 | NP_001268668.1 | 9 | 974 | ||
NM_001281740.2 | NP_001268669.1 | 9 | 974 | |||
NM_025135.4 | NP_079411.2 | 9 | 974 | |||
XM_005258349.1 | XP_005258406.1 | 9 | 956 | |||
XM_005258354.1 | XP_005258411.1 | 9 | 956 | |||
XM_005258355.1 | XP_005258412.1 | 9 | 956 | |||
XM_011526189.1 | XP_011524491.1 | 9 | 956 | |||
XM_011526190.1 | XP_011524492.1 | 9 | 956 | |||
XM_011526192.1 | XP_011524494.1 | 9 | 956 | |||
XM_011526193.2 | XP_011524495.1 | 9 | 956 | |||
XM_011526195.1 | XP_011524497.1 | 9 | 956 | |||
XM_011526196.1 | XP_011524498.1 | 9 | 956 | |||
XM_011526197.1 | XP_011524499.1 | 9 | 956 | |||
XM_017026006.1 | XP_016881495.1 | 9 | 956 | |||
XM_017026007.1 | XP_016881496.1 | 7 | 594 | |||
XM_017026008.1 | XP_016881497.1 | 9 | 956 | |||
XM_017026009.1 | XP_016881498.1 | 9 | 956 | |||
XM_017026010.1 | XP_016881499.1 | 7 | 600 | |||
GenBank mRNA | AB051482.1 | - | 3 | 229 | ||
AB084087.1 | - | 9 | 834 | |||
AK091899.1 | - | 5 | 461 | |||
AK128053.1 | - | 9 | 953 | |||
BC031518.1 | - | 5 | 426 | |||
HM191478.1 | - | 9 | 834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
