siRNA Details
Gene Symbol: GGACT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195087.1 | NP_001182016.1 | 3 | 1632 | ||
NM_033110.2 | NP_149101.1 | 2 | 1647 | |||
XM_005254083.2 | XP_005254140.1 | 2 | 1774 | |||
XM_011521129.2 | XP_011519431.1 | 3 | 4610 | |||
XM_017020804.1 | XP_016876293.1 | 3 | 1748 | |||
GenBank mRNA | AK091974.1 | - | 3 | 1320 | ||
AK093228.1 | - | 1 | 860 | |||
AK095850.1 | - | 3 | 1629 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
