siRNA Details
Gene Symbol: GSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017439.3 | NP_059135.2 | 25 | 2016 | ||
XM_011516325.2 | XP_011514627.1 | 25 | 2083 | |||
XM_011516326.2 | XP_011514628.1 | 24 | 2029 | |||
XM_011516327.2 | XP_011514629.1 | 25 | 2083 | |||
XM_011516330.2 | XP_011514632.1 | 18 | 1529 | |||
XM_017012343.1 | XP_016867832.1 | 24 | 1987 | |||
XM_017012344.1 | XP_016867833.1 | 24 | 1983 | |||
XM_017012345.1 | XP_016867834.1 | 25 | 2029 | |||
XM_017012346.1 | XP_016867835.1 | 23 | 1952 | |||
XM_017012347.1 | XP_016867836.1 | 22 | 1815 | |||
XM_017012348.1 | XP_016867837.1 | 26 | 2159 | |||
XM_017012349.1 | XP_016867838.1 | 25 | 2530 | |||
GenBank mRNA | AK026696.1 | - | 3 | 1387 | ||
AK026747.1 | - | 10 | 1745 | |||
AL079277.1 | - | 3 | 179 | |||
BC101499.1 | - | 3 | 145 | |||
BC110797.1 | - | 25 | 2015 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
