siRNA Details
Gene Symbol: GSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017439.3 | NP_059135.2 | 30 | 2541 | ||
XM_011516325.2 | XP_011514627.1 | 30 | 2608 | |||
XM_011516326.2 | XP_011514628.1 | 29 | 2554 | |||
XM_011516330.2 | XP_011514632.1 | 23 | 2054 | |||
XM_017012343.1 | XP_016867832.1 | 29 | 2512 | |||
XM_017012344.1 | XP_016867833.1 | 29 | 2508 | |||
XM_017012346.1 | XP_016867835.1 | 28 | 2477 | |||
XM_017012347.1 | XP_016867836.1 | 27 | 2340 | |||
XM_017012348.1 | XP_016867837.1 | 31 | 2684 | |||
XM_017012349.1 | XP_016867838.1 | 30 | 3055 | |||
GenBank mRNA | AK026696.1 | - | 8 | 1912 | ||
AK026747.1 | - | 15 | 2270 | |||
AK027188.1 | - | 1 | 1330 | |||
AK126364.1 | - | 3 | 2358 | |||
AL079277.1 | - | 8 | 704 | |||
AL079297.1 | - | 6 | 496 | |||
BC101499.1 | - | 8 | 670 | |||
BC110797.1 | - | 30 | 2540 | |||
BC143549.1 | - | 7 | 599 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
