siRNA Details
Gene Symbol: POM121C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099415.2 | NP_001092885.2 | 15 | 5528 | ||
GenBank mRNA | AK125865.1 | - | 1 | 3390 | ||
AK127026.1 | - | 2 | 3746 | |||
AL050158.1 | - | 1 | 1226 | |||
BC013954.2 | - | 1 | 1044 | |||
BC082993.1 | - | 15 | 5505 | |||
BX649044.1 | - | 2 | 2101 |
Gene Symbol: NSUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EC442348.1 | - | 1 | 21 |
Gene Symbol: POM121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172020.4 | NP_742017.1 | 14 | 5148 | ||
XM_005250726.3 | XP_005250783.1 | 13 | 5502 | |||
XM_005250727.3 | XP_005250784.1 | 12 | 5325 | |||
XM_006716196.3 | XP_006716259.1 | 16 | 5763 | |||
XM_011516730.1 | XP_011515032.1 | 10 | 5214 | |||
XM_017012853.1 | XP_016868342.1 | 16 | 5751 | |||
XM_017012854.1 | XP_016868343.1 | 16 | 5391 | |||
GenBank mRNA | AL080109.1 | - | 3 | 2040 | ||
AL713666.1 | - | 3 | 2041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
