siRNA Details
Gene Symbol: CEP162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286206.1 | NP_001273135.1 | 6 | 609 | ||
NM_014895.3 | NP_055710.2 | 6 | 663 | |||
XM_005248674.3 | XP_005248731.1 | 5 | 786 | |||
XM_005248678.3 | XP_005248735.1 | 6 | 917 | |||
XM_006715380.2 | XP_006715443.1 | 7 | 757 | |||
XM_011535594.2 | XP_011533896.1 | 6 | 917 | |||
XM_017010483.1 | XP_016865972.1 | 5 | 1342 | |||
XM_017010484.1 | XP_016865973.1 | 7 | 960 | |||
GenBank mRNA | AB023226.1 | - | 6 | 570 | ||
AK308190.1 | - | 6 | 630 | |||
BC063632.1 | - | 6 | 636 | |||
DC424230.1 | - | 6 | 645 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
