siRNA Details
Gene Symbol: CEP162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286206.1 | NP_001273135.1 | 13 | 1709 | ||
NM_014895.3 | NP_055710.2 | 13 | 1763 | |||
XM_005248674.3 | XP_005248731.1 | 12 | 1886 | |||
XM_005248678.3 | XP_005248735.1 | 13 | 2017 | |||
XM_006715380.2 | XP_006715443.1 | 14 | 1857 | |||
XM_011535592.2 | XP_011533894.1 | 3 | 617 | |||
XM_011535594.2 | XP_011533896.1 | 13 | 2017 | |||
XM_017010483.1 | XP_016865972.1 | 12 | 2442 | |||
XM_017010484.1 | XP_016865973.1 | 14 | 2060 | |||
GenBank mRNA | AB023226.1 | - | 13 | 1670 | ||
AK023613.1 | - | 4 | 770 | |||
AK308190.1 | - | 13 | 1730 | |||
BC063632.1 | - | 13 | 1736 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
