siRNA Details
Gene Symbol: CEP162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286206.1 | NP_001273135.1 | 19 | 2517 | ||
NM_014895.3 | NP_055710.2 | 19 | 2571 | |||
XM_005248674.3 | XP_005248731.1 | 18 | 2694 | |||
XM_006715380.2 | XP_006715443.1 | 20 | 2665 | |||
XM_011535592.2 | XP_011533894.1 | 9 | 1425 | |||
XM_017010483.1 | XP_016865972.1 | 18 | 3250 | |||
XM_017010484.1 | XP_016865973.1 | 20 | 2868 | |||
GenBank mRNA | AB023226.1 | - | 19 | 2478 | ||
AK023613.1 | - | 10 | 1582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
