siRNA Details
Gene Symbol: URB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314021.1 | NP_001300950.1 | 4 | 1595 | ||
NM_014777.3 | NP_055592.2 | 4 | 1564 | |||
XM_005273360.3 | XP_005273417.1 | 4 | 1533 | |||
GenBank mRNA | AK291013.1 | - | 4 | 1564 | ||
BC059381.1 | - | 4 | 1539 | |||
BC114559.1 | - | 4 | 1517 | 40082897 | ||
BC114963.1 | - | 4 | 1486 | 40082900 | ||
D50923.2 | - | 4 | 1546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
