siRNA Details
Gene Symbol: KIAA1614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020950.1 | NP_066001.1 | 7 | 3214 | ||
GenBank mRNA | AB046834.1 | - | 7 | 3202 | ||
AK090668.1 | - | 4 | 2369 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
