siRNA Details
Gene Symbol: DOCK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290263.1 | NP_001277192.1 | 2 | 281 | ||
NM_014689.2 | NP_055504.2 | 2 | 424 | |||
XM_005246673.4 | XP_005246730.1 | 2 | 283 | |||
XM_006712617.3 | XP_006712680.1 | 2 | 448 | |||
XM_017004441.1 | XP_016859930.1 | 2 | 451 | |||
XM_017004442.1 | XP_016859931.1 | 2 | 454 | |||
XM_017004443.1 | XP_016859932.1 | 2 | 500 | |||
XM_017004444.1 | XP_016859933.1 | 2 | 452 | |||
XM_017004445.1 | XP_016859934.1 | 2 | 281 | |||
XM_017004446.1 | XP_016859935.1 | 2 | 452 | |||
XM_017004447.1 | XP_016859936.1 | 2 | 450 | |||
GenBank mRNA | AB014594.2 | - | 2 | 424 | ||
DB128847.1 | - | 2 | 281 | |||
EU236710.2 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
