siRNA Details
Gene Symbol: ALDH1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034173.3 | NP_001029345.2 | 4 | 675 | ||
NR_027752.1 | 4 | 675 | ||||
XM_011537986.2 | XP_011536288.1 | 2 | 234 | |||
XM_011537988.2 | XP_011536290.1 | 4 | 696 | |||
XM_011537989.2 | XP_011536291.1 | 4 | 696 | |||
XM_017018889.1 | XP_016874378.1 | 5 | 881 | |||
XM_017018890.1 | XP_016874379.1 | 4 | 696 | |||
GenBank mRNA | AK095827.1 | - | 4 | 570 | ||
AK300373.1 | - | 4 | 570 | |||
BC103934.1 | - | 4 | 570 | 40008174 | ||
BC103935.1 | - | 4 | 570 | 40008176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
