siRNA Details
Gene Symbol: TBC1D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286658.2 | NP_001273587.1 | 14 | 2992 | ||
NM_001286659.2 | NP_001273588.1 | 13 | 2827 | |||
NM_014832.4 | NP_055647.2 | 15 | 3016 | |||
XM_005266603.1 | XP_005266660.1 | 14 | 3419 | |||
XM_005266605.2 | XP_005266662.1 | 15 | 2344 | |||
XM_006719903.3 | XP_006719966.1 | 15 | 2704 | |||
XM_011535331.1 | XP_011533633.1 | 14 | 3383 | |||
XM_017020882.1 | XP_016876371.1 | 13 | 2036 | |||
XM_017020883.1 | XP_016876372.1 | 12 | 2030 | |||
XM_017020884.1 | XP_016876373.1 | 3 | 1602 | |||
GenBank mRNA | AB011175.1 | - | 16 | 2965 | ||
AB449885.1 | - | 16 | 2618 | |||
AK300468.1 | - | 4 | 525 | |||
AY211910.1 | - | 4 | 587 | |||
BC151239.1 | - | 16 | 2957 | 8860512 | ||
CR749622.1 | - | 5 | 796 | |||
FM207106.1 | - | 13 | 2426 | |||
FM207107.1 | - | 14 | 2591 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
