siRNA Details
Gene Symbol: NACAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146334.1 | NP_001139806.1 | 2 | 3130 | ||
XM_006715674.3 | XP_006715737.1 | 2 | 3679 | |||
GenBank mRNA | AB002361.1 | - | 1 | 3012 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
