siRNA Details
Gene Symbol: ZFHX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033400.2 | NP_207646.2 | 10 | 8711 | ||
XM_005268135.3 | XP_005268192.1 | 6 | 5874 | |||
XM_011537245.2 | XP_011535547.1 | 10 | 8631 | |||
XM_011537246.2 | XP_011535548.1 | 10 | 11067 | |||
XM_011537247.2 | XP_011535549.1 | 10 | 9646 | |||
XM_011537252.2 | XP_011535554.1 | 6 | 5849 | |||
XM_017021714.1 | XP_016877203.1 | 10 | 8626 | |||
XM_017021715.1 | XP_016877204.1 | 10 | 8553 | |||
XM_017021716.1 | XP_016877205.1 | 9 | 8391 | |||
XM_017021718.1 | XP_016877207.1 | 7 | 5650 | |||
GenBank mRNA | AB051549.1 | - | 2 | 5019 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
