siRNA Details
Gene Symbol: ABHD17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021214.1 | NP_067037.1 | 3 | 1234 | ||
GenBank mRNA | AK091780.1 | - | 1 | 1211 | ||
AK092758.1 | - | 3 | 1312 | |||
AK298606.1 | - | 3 | 820 | |||
AL390079.1 | - | 3 | 855 | |||
BC059401.1 | - | 3 | 1204 | 30346383 | ||
BC115003.1 | - | 3 | 1132 | 7503983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
