siRNA Details
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009959.2 | NP_001009959.1 | 2 | 343 | ||
NM_001304344.1 | NP_001291273.1 | 2 | 191 | |||
NM_001304345.1 | NP_001291274.1 | 2 | 203 | |||
NM_001304346.1 | NP_001291275.1 | 1 | 376 | |||
NM_020711.2 | NP_065762.1 | 1 | 376 | |||
GenBank mRNA | AB033015.1 | - | 1 | 118 | ||
AI207881.1 | - | 1 | 351 | |||
AK295051.1 | - | 1 | 252 | |||
AK295844.1 | - | 1 | 252 | |||
AK296477.1 | - | 2 | 286 | |||
AV727436.1 | - | 2 | 264 | |||
BC026345.1 | - | 1 | 202 | 4798869 | ||
BI598234.1 | - | 1 | 332 | |||
DA248817.1 | - | 2 | 203 | |||
DA288720.1 | - | 2 | 130 | |||
DC322056.1 | - | 2 | 344 | |||
DC331726.1 | - | 1 | 377 | |||
DC343891.1 | - | 1 | 362 | |||
DQ334271.1 | - | 1 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
