siRNA Details
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009959.2 | NP_001009959.1 | 4 | 1062 | ||
NM_001304344.1 | NP_001291273.1 | 4 | 910 | |||
NM_001304345.1 | NP_001291274.1 | 4 | 922 | |||
NM_001304346.1 | NP_001291275.1 | 3 | 1035 | |||
NM_020711.2 | NP_065762.1 | 3 | 1095 | |||
GenBank mRNA | AB033015.1 | - | 3 | 837 | ||
AK295051.1 | - | 2 | 770 | |||
AK295844.1 | - | 3 | 911 | |||
AK296477.1 | - | 4 | 1005 | |||
AK296489.1 | - | 3 | 656 | |||
BC026345.1 | - | 3 | 921 | 4798869 | ||
DQ334271.1 | - | 3 | 918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
