siRNA Details
Gene Symbol: ERMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009959.2 | NP_001009959.1 | 4 | 1815 | ||
NM_001304344.1 | NP_001291273.1 | 4 | 1663 | |||
NM_001304345.1 | NP_001291274.1 | 4 | 1675 | |||
NM_001304346.1 | NP_001291275.1 | 3 | 1788 | |||
NM_020711.2 | NP_065762.1 | 3 | 1848 | |||
GenBank mRNA | AB033015.1 | - | 3 | 1590 | ||
BC026345.1 | - | 3 | 1674 | 4798869 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
