siRNA Details
Gene Symbol: NUP188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015354.2 | NP_056169.1 | 9 | 720 | ||
GenBank mRNA | BX537774.1 | - | 6 | 364 | ||
D79991.1 | - | 9 | 682 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
