siRNA Details
Gene Symbol: NUP188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015354.2 | NP_056169.1 | 43 | 4990 | ||
GenBank mRNA | AK025292.1 | - | 10 | 1315 | ||
BC040352.1 | - | 30 | 3597 | |||
BC111045.1 | - | 21 | 2639 | |||
BX537774.1 | - | 40 | 4634 | |||
D79991.1 | - | 43 | 4952 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
