siRNA Details
Gene Symbol: FRRS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014334.2 | NP_055149.2 | 4 | 689 | ||
XM_011518453.2 | XP_011516755.1 | 4 | 502 | |||
XM_011518454.2 | XP_011516756.1 | 5 | 1073 | |||
GenBank mRNA | AF155065.1 | - | 4 | 689 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
